Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ IDO Mouse anti-Human, Clone: 1F8.2
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MAB10009
Description
Anti-IDO Antibody, clone 1F8.2 is an antibody against IDO for use in western blot, immunohistochemistry.
Specifications
IDO | |
Monoclonal | |
Unconjugated | |
NP_002155.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for up to 12 months. | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
1F8.2 | |
Mouse monoclonal in buffer containing 0.1MTris-glycine, pH 7.4, 150mM NaCl and 0.05% sodium azide. | |
CD107B , EC 1.13.11.52 , IDO, INDO | |
GST-tagged recombinant protein of full-length human IDO. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction