Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IDO (Indoleamine 2, 3-Dioxygenase) Mouse anti-Human, Mouse, Clone: 10.1, MilliporeSigma™

Catalog No. 05840MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-840-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-840-MI Supplier MilliporeSigma Supplier No. 05840
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IDO (Indoleamine 2, 3-Dioxygenase) Antibody, clone 10.1 is an antibody against IDO (Indoleamine 2 for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IDO (Indoleamine 2
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 10.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG in buffer containing 70% storage buffer 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide and 30% glycerol.
Gene Accession No. NP_002155
Gene Symbols INDO, IDO
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 78-184 of human Indoleamine 2,3- Dioxygenase (IDO). Clone 10.1.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.