Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

interferon, alpha-inducible protein 27, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017753 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-753 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-753 Supplier Abnova Corporation Supplier No. H00003429D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human IFI27 protein.

Sequence: MEASALTSSAVTSVAKVVRVASGSAVVLPLARIATVVIGGVVAVPMVLSAMGFTAAGIASSSIAAKMMSAAAIANGGGVASGSLVATLQSLGATGLSGLTKFILGSIGSAIAAVIARFY

Specifications

Antigen interferon, alpha-inducible protein 27
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human IFI27 protein.
Formulation PBS with no preservative; pH 7.4
Gene IFI27
Gene Accession No. NM_005532.3
Gene Alias FAM14D/ISG12/ISG12A/P27
Gene Symbols IFI27
Host Species Rabbit
Immunogen IFI27 (NP_005523.3, 1 a.a. ∼ 119 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3429
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.