Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IFIT1/p56 Rabbit anti-Mouse, Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABF117
Description
Detect IFIT1 using this rabbit polyclonal antibody, Anti-IFIT1/p56 Antibody validated for use in western blotting, immunocytochemistry and Flow Cytometry.
Specifications
IFIT1/p56 | |
Polyclonal | |
Rabbit polyclonal serum containing 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Human, Mouse | |
Serum |
Flow Cytometry, Immunocytochemistry, Western Blot | |
Unconjugated | |
Ifit1; Garg16; Ifi56; Isg56 | |
GST-tagged recombinant protein corresponding to mouse IFIT1/p56. | |
100 μL | |
NP_032357 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction