Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-a/b Receptor 1 Mouse anti-Mouse, NA/LE, Unlabeled, Clone: MAR1-5A3, BD

Catalog No. BDB561183 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561183 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561183 Supplier BD Biosciences Supplier No. 561183
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MAR1-5A3 monoclonal antibody specifically binds to the IFN-α/β Receptor 1 (also known as Interferon-alpha/beta Receptor 1; Type I Interferon Receptor 1 or IFNAR-1) subunit. A variety of cell types including lymphocytes, monocytes/macrophages, dendritic cells, and fibroblasts can be stimulated or induced by viral and microbial infections to produce and secrete type I interferons, including Interferon-alpha (IFN-α) subtypes and Interferon-beta (IFN- β). IFN-α subtypes and IFN-β bind to a common heterodimeric receptor complex (also known as the Type I IFN Receptor or IFN-α/β Receptor) that is comprised of transmembrane glycoprotein IFNAR-1 and IFNR-2 subunits and is expressed on most cell types. Upon ligand binding, IFNAR-1 and IFNAR-2 signal cellular responses. Type I interferons are multifunctional proteins that can induce antiviral states in cells as well as regulate the activation, growth, proliferation, differentiation and viability of various cell types including cells that mediate innate and adaptive immunity as well as autoimmune diseases. The MAR1-5A3 monoclonal antibody blocks Type I IFN receptor signaling and in vitro and in vivo biologic responses caused by type I interferons. The MAR1-5A3 antibody has also been reported to be useful for immunofluorescent staining and flow cytometric analysis, immunoprecipitation and Western blot analysis of IFNAR-1.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-a/b Receptor 1
Applications Flow Cytometry
Classification Monoclonal
Clone MAR1-5A3
Concentration 1.0mg/mL
Conjugate Unconjugated
Description Interferon-alpha/beta Receptor 1; IFN-R-1; Ifar; Ifnar; Ifrc; Infar
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Mouse
Immunogen Mouse Ifnar1 extracellular domain DNA
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.