Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-g Rat anti-Mouse, Alexa Fluor 488, Clone: XMG1.2, BD

Catalog No. BDB557724 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557724 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557724 Supplier BD Biosciences Supplier No. 557724
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The XMG1.2 antibody reacts with mouse interferon-γ (IFN-γ) protein. IFN-γ is a pleiotropic cytokine, of approximately 15-17kDa, involved in the regulation of the immune response. It plays an important role in activation, growth, and differentiation of T and B lymphocytes, macrophages, NK cells and other non-hematopoietic cell types. IFN-γ production is associated with the Th-1 differentiation. This is a neutralizing antibody

Host Species: Rat
Clone: XMG1.2
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse IFN-<special id='307'/>

Flow Cytometry, Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-g
Applications Flow Cytometry
Classification Monoclonal
Clone XMG1.2
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse IFN-γ Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.