Learn More
IFN-gamma Mouse anti-Human, Brilliant Violet 650, Clone: 4S.B3, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 563416
Description
The 4S.B3 monoclonal antibody specifically binds to interferon-γ(IFN-γ). The immunogen used to generate this hybridoma was partially purified human IFN-γobtained from supernatants of human PBMC stimulated with Staphylococcus aureus . Interferon-γ(IFN-γ) is a potent multifunctional cytokine that is produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ + T cells. IFN- γexerts its biological effects through specific binding to the high-affinity IFN-γReceptor Complex comprised of IFN-γRα (CD119) and ese influences can involve IFN- γ’s capacity to boost MHC class I and II expression by antigen-presenting cells as well as to direct effects on B cells and T cells themselves. Human IFN-γis a 14- 18 kDa glycoprotein containing 143 amino acid residues.

Specifications
IFN-gamma | |
Monoclonal | |
Brilliant Violet 650 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Partially purified human IFN-gamma from supernatants of human PBMC stimulated with Staphylococcus aureus | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
4S.B3 | |
IFNG; Interferon-gamma; IFG; IFI; Type II interferon | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.