Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-gamma Mouse anti-Human, Brilliant Violet 786, Clone: 4S.B3, BD

Catalog No. BDB563731 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563731 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563731 Supplier BD Biosciences Supplier No. 563731
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 4S.B3 monoclonal antibody specifically binds to interferon-γ(IFN-γ). The immunogen used to generate this hybridoma was partially purified human IFN-γobtained from supernatants of human PBMC stimulated with Staphylococcus aureus. Interferon-γ(IFN-γ) is a potent multifunctional cytokine that is produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ+ T cells. IFN-γexerts its biological effects through specific binding to the high-affinity IFN-γReceptor Complex comprised of IFN-γRα (CD119) and ese influences can involve IFN-γ’s capacity to boost MHC class I and II expression by antigen-presenting cells as well as to direct effects on B cells and T cells themselves. Human IFN-γis a 14-18 kDa glycoprotein containing 143 amino acid residues.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-gamma
Applications Flow Cytometry
Classification Monoclonal
Clone 4S.B3
Conjugate Brilliant Violet 786
Description IFNG; Interferon-gamma; IFG; IFI; Type II interferon
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Partially purified human IFN-γ from supernatants of human PBMC stimulated with Staphylococcus aureus
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.