Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-g Hamster anti-Mouse, NA/LE, Unlabeled, Clone: H22, BD

Catalog No. BDB557530 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557530 0.25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557530 Supplier BD Biosciences Supplier No. 557530
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The H22 antibody reacts with mouse interferon-γ (IFN-γ , but not mouse IFN-α , IFN-β , IL-6, or human IFN-γ. The H22 antibody was reported to inhibit the in vitro anti-viral, the class II MHC antigen-inducing and the macrophage-activating factor (MAF) activities of mouse IFN-γ. Moreover, the IFN-γ neutralizing capacity of has been reported in several in vivo model systems. The specific binding of I[125]-labeled recombinant mouse IFN-γ to IFN-γ receptor-positive cells is blocked by H22. Although the H22 antibody is reported to react with rat IFN-γ , its reactivity with IFN-γ from other species has not been tested.

Host Species: Hamster
Clone: H22
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Recombinant Mouse IFN-<special id='307'/>

Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-g
Applications ELISA
Classification Monoclonal
Clone H22
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Host Species Hamster
Immunogen Recombinant Mouse IFN-γ
Purification Method Affinity Purified
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.