Learn More
Description
Host Species: Rat
Clone: XMG1.2
Isotype: IgG1, κ
Species Reactivity: Ms
Immunogen: Mouse IFN-γ Recombinant Protein
Flow Cytometry (Intracellular Staining)
Specifications
Specifications
| Antigen | IFN-gamma |
| Classification | Monoclonal |
| Clone | XMG1.2 |
| Concentration | 0.2mg/mL |
| Host Species | Rat |
| Immunogen | Mouse IFN-γ Recombinant Protein |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.