Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IFN gamma, APC, Clone: XMG1.2,

Catalog No. MABF1516MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1516MI 100 μg
MABF1515MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1516MI Supplier MilliporeSigma™ Supplier No. MABF1516
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects IFN gamma Clone: XMG1.2 in Mouse samples, and it is validated for Flow Cytometry

The XMG1.2 antibody is specific for mouse Interferon-gamma (IFN-g), a 20kDa type II cytokine known for its central roles in protection against bacterial or viral pathogens and for its anti-tumor properties. IFN-g is secreted by several types of immune cells, which allow the cytokine to modulate innate immunity, when secreted by NK and NKT cells, and to function in support of adaptive immunity when secreted by Th1 and CD8+ T cells (CTLs).The XMG1.2 antibody is suitable for detection of intracellular IFN-g protein, e.g. by flow cytometry, as well as for quantitative analysis of the secreted protein by ELISA, when paired with an appropriate secondary antibody. This clone is also widely used for neutralization of the functional activity of IFN-g in a variety of assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN gamma
Applications Flow Cytometry
Classification Monoclonal
Clone XMG1.2
Conjugate APC
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P01580
Gene Symbols Ifng
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.