Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-g Mouse anti-Human, PE, Clone: 4S.B3, BD

Catalog No. BDB557074 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557074 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557074 Supplier BD Biosciences Supplier No. 557074
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone 4S.B3 reacts with the human form of interferon-γ (IFN-γ) detectable in the cytoplasm of a major subset of activated peripheral blood T lymphocytes. Clone 4S.B3 also cross-reacts with a cytoplasmic component of peripheral blood CD3+ lymphocytes of baboon, and both rhesus and cynomolgus macaque monkeys following five-hour treatment with phorbol myristic acetate (PMA) and Ca++ ionophore (A23187) in the presence of monensin. The staining pattern of 4S.B3 in CD3+ cells is similar to that observed with peripheral blood T lymphocytes from normal human donors. This reagent is useful for intracellular immunofluorescent staining for flow cytometric analysis to identify and enumerate IFN-γ + cells within a mixed cell population. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 4S.B3
Isotype: IgG1, κ
Species Reactivity: Human, Baboon/Rhesus/Cynomolgus
Immunogen: Partially purified human IFN-<special id='307'/> from supernatants of human PBMC stimulated with Staphylococcus aureus

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-g
Applications Flow Cytometry
Classification Monoclonal
Clone 4S.B3
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IFN-γ from supernatants of S. aureus-stimulated PBMC
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.