Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ Mouse anti-Human, PerCP-Cy5.5, Clone: B27, BD

Catalog No. BDB560704 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560704 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560704 Supplier BD Biosciences Supplier No. 560704
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The B27 monoclonal antibody specifically binds to human interferon- γ(IFN- γ). IFN-γis a potent multifunctional cytokine that is produced by several activated cell types including NK, NKT, CD4+TCRαβ+, CD8+TCRαβ+, and TCRγδ + T cells. IFN- γexerts i ts biological effects through specific binding to the high-affinity IFN- γReceptor Complex comprised of IFN- γβ subunits. In addition to its antiviral effects, IFN- γupregulates a number of lymphoid cell functions including the antimicrobial and antitumor responses of macrophages, NK cells, and neutrophils. In addition, IFN- γcan exert strong regulatory influences on the proliferation, differentiation, and effector responses of B cell and T cell subsets. Th ese influences can involve IFN- γ’s capacity to boost MHC class I and II expression by antigen-presenting cells as well as to direct effects on B cells and T cells t hemselves. Human IFN-γis a 14- 18 kDa glycoprotein containing 143 amino acid residues. B27 is a neutralizing antibody. The use of B27 antibody for epitope mapping of human IFN- γhas been described. The B27 antibody has been reported not to bind to denatured IFN- γ.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry
Classification Monoclonal
Clone B27
Conjugate PerCP-Cyanine5.5
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human IFN-γ Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.