Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ Mouse anti-Pig, PerCP-Cy5.5, Clone: P2G10, BD

Catalog No. BDB561481 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561481 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561481 Supplier BD Biosciences Supplier No. 561481
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The P2G10 monoclonal antibody specifically binds to porcine interferon-γ (IFN-γ). The immunogen used to generate the P2G10 hybridoma was recombinant pig IFN-γ protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Flow Cytometry
Classification Monoclonal
Clone P2G10
Concentration 0.2mg/mL
Conjugate PerCP-Cyanine5.5
Description IFNG; IFN-gamma; Interferon gamma; Interferon-gamma
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant pig IFN-γ protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.