Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFN-γ Mouse anti-Rat, PE, Clone: DB-1, BD

Catalog No. BDB559499 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559499 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559499 Supplier BD Biosciences Supplier No. 559499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DB-1 antibody reacts with rat interferon-γ (IFN-γ). The immunogen used to generate the DB-1 hybridoma was CHO cell line expressed recombinant rat IFN-γ. This is a neutralizing antibody.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen IFN-γ
Applications Blocking Assay, Flow Cytometry
Classification Monoclonal
Clone DB-1
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Rat IFN-γ
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.