Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IFNAR-1 Mouse anti-Mouse, Clone: MARI-5A3, MilliporeSigma™

Catalog No. 04151MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-151-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-151-MI Supplier MilliporeSigma Supplier No. 04151
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IFNAR-1 Antibody, clone MARI-5A3 is an antibody against IFNAR-1 for use in flow cytometry, immunoprecipitation, western blot, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IFNAR-1
Applications Flow Cytometry, Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone MARI-5A3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_034638
Gene Symbols IFRC, IFNAR1, IFNBR, IFN-R-1, IFNAR, AVP, CRF2-1
Host Species Mouse
Immunogen Recombinant protein corresponding to the extracellular domain of mouse IFNAR-1.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.