Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig Goat anti-Mouse, Brilliant Violet 421, Polyclonal, BD

Catalog No. BDB563846 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563846 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563846 Supplier BD Biosciences Supplier No. 563846
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BD Horizon™ BV421 Goat Anti-Mouse Ig is intended to be a second-step reagent for staining of cells pre-stained with primary mouse Ig antibodies. It is reactive with the heavy and light chains of mouse primary antibodies having the IgG1, IgG2a, IgG2b, IgG3, IgM, and IgA isotypes. Minimal background staining of human and rat leucocytes occurs in the absence of a primary mouse antibody. In addition, the BD Horizon BV421 Goat Anti-Mouse Ig stains mouse peripheral B lymphocytes with little non-specific staining of other splenic leucocytes. Therefore, it is useful as a primary reagent in immunofluorescent staining of mouse B cells and antibody-producing cells and as a secondary reagent for staining cells pre-stained with mouse Ig primary antibodies.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Goat
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.