Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig Goat anti-Rat, Brilliant Violet 421, Polyclonal, BD

Catalog No. BDB565013 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565013 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565013 Supplier BD Biosciences Supplier No. 565013
Add to Cart
Add to Cart

Goat Polyclonal Antibody

BD Horizon™ BV421 Goat Anti-Rat Ig is intended to be a second-step reagent for immunofluorescent staining of cells pre-stained with Rat Ig primary antibodies. It also stains rat B cells and has little reactivity with rat non-B cells or mouse splenocytes. As a second step, it is reactive with rat IgG and IgM monoclonal antibodies although a weaker signal may be detected when the primary antibody has a rat IgM or IgG2b isotype due to its adsorption with mouse Ig. It has weak crossreactivity detectable by flow cytometry with some, but not all, hamster immunoglobulins.

The antibody was conjugated to BD Horizon BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. BD Horizon BV421 has an Ex Max at 407nm and Em Max at 421 nm. The use of a mounting reagent (eg, ProLong™ Gold) is highly recommended to maximize the photostability of BV421.

For epifluorescence microscopes with broad spectrum excitation sources, the recommended excitation and emission filters are 392/23nm and 430/24nm bandpass filters, respectively. For specific multicolor imaging applications, the exact filter configurations should be optimized by the end user. For additional instrument/filter configuration information, please visit http://www.bdbiosciences.com/research/cellularimaging.

For flow cytometry, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Goat
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.