Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig,κ Light Chain Rat anti-Mouse, Brilliant Violet 421, Clone: 187.1, BD

Catalog No. BDB562888 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562888 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562888 Supplier BD Biosciences Supplier No. 562888
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse λ1 or λ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407nm and Em Max at 421nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.

Host Species: Rat
Clone: 187.1
Isotype: IgG1, κ
Species Reactivity: Ms
Immunogen: Mouse IgG2b ? secreted by MPC-11 plasmacytoma

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig,κ Light Chain
Applications Flow Cytometry
Classification Monoclonal
Clone 187.1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 421
Description Ig kappa chain C region; IGKC; Igk-C; Ig, κ
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen IgG2b ? secreted by MPC-11 plasmacytoma
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.