Learn More
Kappa, Ig light chain Mouse anti-Human, Unlabeled, Clone: TB28-2, BD
Shop All BD Biosciences ProductsDescription
Anti-Kappa is specific for kappa light chains of human immunoglobulins. Immunoglobulins bearing kappa light chains are present on approximately 60% of normal B lymphocytes and on Igκ+ neoplastic cells. In serum, Anti-Kappa reacts with immunoglobulins bearing kappa light chains as well as free kappa light chains.
Host Species: Mouse
Clone: TB28-2
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | Kappa, Ig light chain |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | TB28-2 |
| Concentration | 12.5μg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human IgG, κ myeloma protein |
| Purification Method | Affinity Purified |
| Show More |
Safety and Handling
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.