Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig, κ light chain Rat anti-Mouse, HRP, Clone: 187.1, BD

Catalog No. BDB559751 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559751 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559751 Supplier BD Biosciences Supplier No. 559751

Rat Monoclonal Antibody

The 187.1 antibody reacts specifically with mouse Igs bearing κ light chain. It does not react with λ1 or λ2 light chain or heavy chain.

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig, κ light chain
Applications ELISA
Classification Monoclonal
Clone 187.1
Conjugate HRP
Description Ig kappa chain C region; IGKC; Igk-C; Ig, κ
Formulation Aqueous buffered solution containing BSA and ProClin™150 as preservative.
Host Species Rat
Immunogen IgG2b κ secreted by MPC-11 plasmacytoma
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.