Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ig κ Light Chain Rat anti-Mouse, PE, Clone: 187.1, BD
Rat Monoclonal Antibody
$128.00 - $234.00
Specifications
Antigen | Ig κ Light Chain |
---|---|
Clone | 187.1 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB562021
|
BD Biosciences
562021 |
25 μg |
Each for $128.00
|
|
|||||
BDB559940
|
BD Biosciences
559940 |
0.1 mg |
Each for $234.00
|
|
|||||
Description
The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse μ1 or μ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.
Specifications
Ig κ Light Chain | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
IgG2b κ secreted by MPC-11 plasmacytoma | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
187.1 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title