Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kappa, Ig light chain Rat anti-Mouse, PE, Clone: X36, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 349073
Description
Rat Anti-Mouse Kappa, clone X36, is derived from hybridization of rat YB2/0 myeloma cells with spleen cells from LOU/M rats immunized with mouse IgM,κ.

Specifications
Kappa, Ig light chain | |
Monoclonal | |
12.5μg/mL | |
Buffered saline with gelatin and 0.1% sodium azide. | |
Mouse IgM, κ | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
X36 | |
PE | |
Rat | |
100 Tests | |
Immunology | |
Mouse | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction