Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ig, κ Rat anti-Mouse, PE-Cy7, Clone: 187.1, BD

Catalog No. BDB560667 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560667 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560667 Supplier BD Biosciences Supplier No. 560667
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 187.1 monoclonal antibody specifically binds to kappa light chains of mouse immunoglobulins. The 187.1 antibody does not react with mouse μ1 or μ2 immunoglobulin lights chains or mouse immunoglobulin heavy chains.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Ig, κ
Applications Flow Cytometry
Classification Monoclonal
Clone 187.1
Concentration 0.2mg/mL
Conjugate PE-Cyanine7
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen IgG2b κ secreted by MPC-11 plasmacytoma
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.