Learn More
IgA Rat anti-Mouse, Biotin, Clone: C10-1, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 556978
Description
The C10-1 antibody reacts specifically with mouse IgA of Igh-C[a] and Igh-C[b] haplotypes. It does not react with other Ig isotypes. Detection of surface immunoglobulin on B lymphoma cells has been demonstrated with C10-1 mAb.
Host Species: Rat
Clone: C10-1
Isotype: IgG1, κ
Cross Reactivity: Mouse
ELISA, Flow Cytometry

Specifications
IgA | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Pooled mouse IgA | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 κ |
ELISA | |
C10-1 | |
Biotin | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.