Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Mouse anti-Chicken, FITC, Clone: A-1, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 833002
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
IgA | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
0.5 mg | |
Primary | |
Store at 2-8°C; Protect from light |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
A-1 | |
FITC | |
Mouse | |
RUO | |
Chicken | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction