Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgA Rat anti-Mouse, FITC, Clone: C10-3, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559354
Description
The C10-3 antibody reacts specifically with mouse IgA of Igh-C(a) and Igh-C(b) haplotypes. It does not react with other Ig isotypes.
Flow Cytometry, Intracellular Staining

Specifications
IgA | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Pooled mouse IgA | |
0.5 mg | |
Immunology | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
C10-3 | |
FITC | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction