Learn More
Description
Reacts with human immunoglobulin A1 (IgA1). It does not react with human IgA2, nor does it bind to other human immunoglobulin heavy chains. IgA1 is present in serum and secretions and is expressed in less than 1% of normal peripheral blood lymphocytes.
Host Species: Mouse
Clone: CL-155-1
Isotype: IgG3, λ
Species Reactivity: Human
ELISA
Specifications
Specifications
| Antigen | IgA1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | CL-155-1 |
| Concentration | 0.5mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.