Learn More
IgD Mouse anti-Human, APC-H7, Clone: IA6-2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 561305
Description
The IA6-2 monoclonal antibody specifically binds to the heavy chain of human Immunoglobulin D (IgD). IgD is a member of the immunoglobulin superfamily that exists in type 1-membrane (mIgD) and soluble glycoprotein forms. mIgD is expressed on mature naοve B cells (along with membrane IgM) and serves as a B-cell receptor for antigen (BCR). In response to antigen binding, the mIgD BCR, in association with other signaling molecules including CD79a and CD79b, can transduce activating or tolerizing signals intracellularly into B lymphocytes.

Specifications
IgD | |
Monoclonal | |
APC-H7 | |
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
Human IgD | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
IA6-2 | |
IGHD; Ig delta chain C region; Immunoglobulin heavy constant delta | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.