Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgD Rat anti-Mouse, BUV395, Clone: 11-26c.2a, BD

Catalog No. BDB564274 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564274 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564274 Supplier BD Biosciences Supplier No. 564274
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 11-26c.2a monoclonal antibody specifically binds to mouse immunoglobulin D of all Igh-C haplotypes (e.g., IgDa, IgDb, IgDe), and it does not react with other immunoglobulin isotypes. Although 11-26c.2a mAb binds membrane IgD expressed on the splenic B-cell surface with high affinity, it does not induce proliferation of splenic B cells in vitro . In vivo injection of 11-26c.2a antibody does not have any effect on activation of mature B cells, as determined by MHC class II antigen expression.

The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgD
Applications Flow Cytometry
Classification Monoclonal
Clone 11-26c.2a
Concentration 0.2mg/mL
Conjugate Brilliant Ultraviolet 395
Description IGHD; Igh-5; Immunoglobulin heavy chain 5; Ig delta chain C region
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Rat
Immunogen Not reported
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.