Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-I Mouse anti-Human, Mouse, Rat, Chicken, Avian, Clone: Sm1.2, MilliporeSigma™

Catalog No. 05172MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-172-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-172-MI Supplier MilliporeSigma Supplier No. 05172
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IGF-I Antibody, clone Sm1.2 is an antibody against IGF-I for use in immunohistochemistry, immunoprecipitation, NEUT and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-I
Applications Immunohistochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone Sm1.2
Conjugate Unconjugated
Formulation Purified mouse IgG1κ lyophilized powder, lyophilized from 105μL of 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.1mM EDTA.
Gene Accession No. NP_000609
Gene Symbols IGF1, IBP1, IGFI, IGF-IA, MGF, IGF-IB
Host Species Mouse
Immunogen Human insulin-like growth factor I (IGF-I) purified from human plasma. (van Wyk, J.J., 1986).
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Avian, Chicken, Human, Mouse, Rat
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.