Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IGF-II Mouse anti-Human, Rat, Clone: S1F2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05166
Description
Detect IGF-II using this Anti-IGF-II Antibody, clone S1F2 validated for use in ELISA, immunoprecipitation, immunocytochemistry, NEUT and western blotting.
Specifications
IGF-II | |
Monoclonal | |
Unconjugated | |
NM_000612 | |
Mouse | |
Ammonium sulfate precipitation and DEAE-cellulose chromatography | |
Primary | |
−20°C for up to 12 months. | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Neutralization, Western Blot | |
S1F2 | |
Purified mouse monoclonal IgG1 in buffer containing PBS, pH 7.4., purified by ammonium sulfate precipitation and DEAE-cellulose chromatography. Frozen solution. | |
IGF2, FLJ22066, FLJ44734, pp9974, IGF-II, INSIGF, C11orf43 | |
Partially purified rat IGF-II. Clone S1F2 | |
500 μg | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction