Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-IGF-IR Clone: αIR3,

Catalog No. GR11L100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
GR11L100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GR11L100UG Supplier MilliporeSigma™ Supplier No. GR11L100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects IGF-IR Clone: αIR3 in Human samples, and it is validated for Immunofluorescence, Immunoprecipitation

The insulin-like growth factors (IGFs) or somatomedins are mitogenic peptides present in the plasma of human, rat, and other species. Two distinct but chemically related IGFs have been purified from human plasma: IGF-I (Somatomedin C) and IGF-II. They are single chain polypeptides with molecular weights of 7,640 and 7,471, respectively. Types I and II IGF receptors differ in their structure and in their peptide binding specificity. Type I IGF receptors are structurally homologous to insulin receptors and contain α-subunits (Mr=130,000) that bind peptide, and β-subunits (Mr=90,000), which have intrinsic tyrosine kinase activity. The protein-tyrosine kinase activity of the insulin receptor and, presumably the IGF-I receptor, is required for insulin to stimulate all of its biological responses in cells. Type II receptors are a single polypeptide (Mr=260,000), lack protein-tyrosine kinase activity, and have been shown to be identical to the mannose-6-phosphate receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-IR
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone αIR3
Conjugate Unconjugated
Formulation In 0.05M sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen partially purified, human placenta IGF-I receptor
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽130kDa α and the ∽90kDa β subunits of IGF-I receptor.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.