Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF-IR Mouse anti-Human, Mouse, Clone: JBW902, MilliporeSigma™

Catalog No. 05656MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-656-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-656-MI Supplier MilliporeSigma Supplier No. 05656
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IGF-IR Antibody, clone JBW902 detects level of IGF-IR and has been published and validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF-IR
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone JBW902
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.14MTris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide and 30% glycerol.
Gene Accession No. NM_000875
Gene Symbols IGF1R, IGFIR, MGC18216, MGC142170, JTK13, MGC142172, CD221
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 1264-1367 of human IGF-1 Receptor
Purification Method Protein A purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.