Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF1 Receptor (CT) Mouse anti-Human, Clone: 7G11, MilliporeSigma™

Catalog No. 04298MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
04-298-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 04-298-MI Supplier MilliporeSigma Supplier No. 04298
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-IGF1 Receptor (C-terminus) Antibody, clone 7G11 detects level of IGF1 Receptor (C-terminus) and has been published and validated for use in ELISA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF1 Receptor (CT)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7G11
Conjugate Unconjugated
Formulation 100 μg of purified mouse monoclonal IgG lyophilized in 2X PBS containing 0.09% sodium azide, PEG, and sucrose
Gene Symbols IGF1, IBP1, IGFI, IGF-IA, MGF, IGF-IB
Host Species Mouse
Immunogen KLH-conjugated synthetic peptide encompassing the surrounding amino acids of the C-terminus in human IGF1
Purification Method Subsequent thiophilic adsorption and size exclusion chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.