Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGF2BP1/2 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. 07102MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-102-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-102-MI Supplier MilliporeSigma Supplier No. 07102

Rabbit Polyclonal Antibody

Anti-IGF2BP1/2 Antibody is a Rabbit Polyclonal Antibody for detection of IGF2BP1/2 also known as Hepatocellular carcinoma autoantigen p62, IGF2 mRNA-binding protein 2 and has been validated in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGF2BP1/2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal containing PBS with 0.09% azide.
Gene Symbols IGF2BP2; IMP-2; IMP2; VICKZ2; p62; IGF2BP1; IMP2; IMP-1
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human IGF2BP1/2 at the C-terminus.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_006537
Target Species Human, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.