Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IGFBP-1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 06106MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-106-MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-106-MI Supplier MilliporeSigma Supplier No. 06106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect IGFBP-1 using this Anti-IGFBP-1 Antibody validated for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFBP-1
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized
Gene Symbols IGFBP1; IBP1; AFBP; PP12; IGF-BP25; hIGFBP-1; IGFBP-1
Host Species Rabbit
Immunogen IGFBP-1 purified from human amniotic fluid
Purification Method Antiserum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000596
Target Species Human
Content And Storage Lyophilized: 2 years at 4°C Rehydrated: 6 months at −20°C
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.