Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-IgG, Alexa Fluor 700, Polyclonal, R&D Systems™

Catalog No. IC105N
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC105N 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC105N Supplier R&D Systems Supplier No. IC105N
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IgG Polyclonal antibody specifically detects IgG in Rabbit samples. It is validated for Control.

Specifications

Antigen IgG
Applications Control
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Control 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide.
Gene Alias immunoglobulin heavy constant gamma 1 (G1m marker)
Host Species Rabbit
Immunogen NA
Purification Method Protein A or G purified
Quantity 200 Tests
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Serum was obtained from naive (non-immunized) rabbits.
Target Species Rabbit
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.