Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Mouse anti-Human, Brilliant Violet 421, Clone: G18-145, BD

Catalog No. BDB562581 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562581 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562581 Supplier BD Biosciences Supplier No. 562581
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Antibody was conjugated with Brilliant Violet 421 under optimum conditions, and unconjugated antibody and free Brilliant Violet 412 were removed. G18-145 antibody does not reacts with heavy chains of other human immunoglobulin isotypes.

Host Species: Mouse
Clone: G18-145
Isotype: IgG1κ
Species Reactivity: Human

  • With Ex Max of 407nm and Em Max at 421nm, Brilliant Violet 421 can be excited by the violet laser and detected in the standard Pacific Blue filter set
  • Brilliant Violet 421 conjugates are very bright, often exhibiting 10 fold improvement in brightness compared to Pacific Blue conjugates

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Flow Cytometry
Classification Monoclonal
Clone G18-145
Concentration 5μL
Conjugate Brilliant Violet 421
Description IGHG1, IGHG2, IGHG3 and IGHG4
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.