Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Mouse anti-Pig, Unlabeled, Clone: F007-1241, BD

Catalog No. BDB552554 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552554 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552554 Supplier BD Biosciences Supplier No. 552554
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The F007-1241 antibody reacts with pig IgG, but not with pig IgM. The reactivity with individual pig IgG subclasses (IgG1, IgG2a, IgG2b, IgG3, IgG4) was not determined. It does not cross-react with mouse, rat, hamster, or rabbit immunoglobulins. The F007-1241 antibody does have some cross-reactivity with dog immunoglobulins.

Host Species: Mouse
Clone: F007-1241
Isotype: IgG1 κ
Species Reactivity: Porcine
Immunogen: Purified pig IgG

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications ELISA
Classification Monoclonal
Clone F007-1241
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified pig IgG
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Pig
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.