Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG, Cocktail Mouse anti-Hamster, Biotin, Clone: G70-204, G94-56, BD

Catalog No. BDB550335 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550335 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550335 Supplier BD Biosciences Supplier No. 550335

Mouse Monoclonal Antibody

This preparation consists of a mixture of two mouse anti-hamster IgG monoclonal antibodies: G70-204 (isotype IgG1,κ), and G94-56 (isotype IgG2b,κ). It reacts with Armenian hamster IgG, groups 1, 2, and 3, and Syrian hamster IgG, group 1, and group 2 monoclonal antibodies. It does not react with hamster IgM antibodies.

Host Species: Hamster
Clone: G70-2
Species Reactivity: Hamster
Immunogen: Pooled Armenian hamster IgG mAb

Immunohistochemistry, Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG, Cocktail
Applications ELISA
Classification Monoclonal
Clone G70-204, G94-56
Concentration 500μg/mL
Conjugate Biotin
Formulation Aqueous buffered solution containing BSA, protein stabilizer, goat serum, and ≤ 0.09% sodium azide.
Host Species Mouse
Immunogen Pooled Armenian hamster IgG mAb
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Hamster
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.