Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG Detector Mouse anti-Rabbit, PE, Clone: C101-359, BD

Catalog No. BDB558553 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558553 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558553 Supplier BD Biosciences Supplier No. 558553
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The PE Mouse anti-Rabbit IgG Detector is useful in verification of the conjugation of rabbit IgG isotype antibodies to the BD CBA Functional Beads. It does not react with rabbit IgM. It does not cross-react with mouse, rat, human or guinea pig immunoglobulins.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG Detector
Applications Flow Cytometry
Classification Monoclonal
Clone C101-359
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rabbit
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.