Learn More
IgG (Multiple Adsorption) Donkey anti-Rabbit, PE, Polyclonal, BD
Donkey Polyclonal Antibody
Supplier: BD Biosciences 558416
Description
The polyclonal F(ab')2 antibody reacts with heavy chains of rabbit IgG and light chains of most rabbit immunoglobulins, and not with non-immunoglobulin proteins of rabbit serum. It has minimal cross-reactivity with human, mouse, rat, bovine, caprine, equine, ovine, chicken, guinea pig, and Syrian hamster serum proteins.
Control, Intracellular Staining

Specifications
IgG (Monkey adsorbed) | |
Polyclonal | |
PE | |
Donkey | |
50 Tests | |
Immunology | |
Rabbit | |
Liquid |
Flow Cytometry | |
POLYCLONAL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rabbit IgG Protein | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.