Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG1 Mouse, FITC, Clone: X40, BD

Catalog No. BDB349041 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB349041 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB349041 Supplier BD Biosciences Supplier No. 349041
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Control, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG1
Applications Flow Cytometry
Classification Monoclonal
Clone X40
Concentration 50μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Keyhole limpet hemocyanin (KLH)
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.