Learn More
IgG1 Rat anti-Mouse, HRP, Clone: X56, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 559626
Description
The X56 antibody reacts specifically with mouse IgG1 of Igh-C(a) and Igh-C(b) haplotypes. It does not react with other Ig isotypes. This antibody is routinely tested by ELISA analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
ELISA

Specifications
IgG1 | |
Monoclonal | |
HRP | |
Aqueous buffered solution containing BSA and ProClin™150 as preservative. | |
Not Reported | |
1 mL | |
Immunology | |
Mouse | |
IgG1 κ |
ELISA | |
X56 | |
Igh-4; Ighg1; Immunoglobulin heavy constant gamma 1 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.