Learn More
anti-IgG1, κ, Mouse, Alexa Fluor™ 488, Clone: MOPC-21, BD
$170.70 - $285.50
Specifications
Clone | MOPC-21 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugate | Alexa Fluor 488 |
Host Species | Mouse |
Target Species | Human |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB557782
|
BD Biosciences
557782 |
50 Tests |
Each for $170.70
|
|
|||||
BDB557721
|
BD Biosciences
557721 |
100 Tests |
Each for $285.50
|
|
|||||
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
MOPC-21 | |
Alexa Fluor 488 | |
Human | |
Affinity Purified |
Flow Cytometry, Western Blot | |
Mouse | |
IgG1 κ | |
Undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.