Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG1, PE, Clone: X40, BD
$212.00 - $243.00
Specifications
Antigen | IgG1 |
---|---|
Clone | X40 |
Classification | Monoclonal |
Conjugate | PE |
Regulatory Status | RUO |
Description
Control, Flow Cytometry
Specifications
IgG1 | |
Monoclonal | |
RUO | |
Primary |
X40 | |
PE | |
Immunology |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title