Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-IgG2, κ, Clone: B81-3, BD
$208.00 - $280.00
Specifications
Clone | B81-3 |
---|---|
Applications | Blocking Assay, Functional Assay, Cytotoxicity Assay |
Conjugate | Unconjugated |
Host Species | Hamster |
Isotype | IgG2 κ |
Description
The B81-3 antibody is specific for keyhole limpet hemocyanin. This antibody is routinely tested by LAL assay for endotoxin levels. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Blocking Assay, Co-Stimulation, Cytotoxicity Assay, Isotype Control
Specifications
B81-3 | |
Unconjugated | |
IgG2 κ | |
Undiluted at 4°C |
Blocking Assay, Functional Assay, Cytotoxicity Assay | |
Hamster | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title