Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2, l Hamster anti-Mouse, Alexa Fluor 700, Clone: HA4/8, BD

Catalog No. BDB557985 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557985 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557985 Supplier BD Biosciences Supplier No. 557985

Hamster Monoclonal Antibody

The Ha4/8 antibody is specific for the keyhole limpet hempcyanin (KLH).

Host Species: Hamster
Clone: Ha4/8
Isotype: IgG1, <special id='310'/>1
Species Reactivity: Mouse
Immunogen: Keyhole limpet hemocyanin

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2, λ
Applications Flow Cytometry
Classification Monoclonal
Clone HA4/8
Concentration 0.2mg/mL
Conjugate Alexa Fluor 700
Description Anti-KLH
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Hamster
Immunogen Keyhole limpet hemocyanin
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2 λ1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.