Learn More
anti-IgG2a κ, Rat , Alexa Fluor™ 647, Clone: R35-95, BD
$219.00 - $247.00
Specifications
Clone | R35-95 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugate | Alexa Fluor 647 |
Host Species | Rat |
Immunogen | Mouse Pooled Immunoglobulin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB557690
|
BD Biosciences
557690 |
0.1 mg |
Each for $219.00
|
|
|||||
BDB557906
|
BD Biosciences
557906 |
100 Tests |
Each for $247.00
|
|
|||||
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a, κImmunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
R35-95 | |
Alexa Fluor 647 | |
Mouse Pooled Immunoglobulin | |
Affinity Purified |
Flow Cytometry, Western Blot | |
Rat | |
IgG2a κ | |
Undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.